accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3O

CAS RN 928-68-7

IUPAC Name: 6-methylheptan-2-one
Synonyms: 2-heptanone, 6-methyl-
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: DPLGXGDPPMLJHN-UHFFFAOYSA-N
SMILES: CC(C)CCCC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)