accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928-97-2

IUPAC Name: (3E)-hex-3-en-1-ol
Synonyms: trans-3-hexenol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UFLHIIWVXFIJGU-ONEGZZNKSA-N
SMILES: CC\C=C\CCO
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)