accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 929-06-6

IUPAC Name: 2-(2-aminoethoxy)ethan-1-ol
Synonyms: 2-(2-aminoethoxy)ethanol
Molecular Weight (g/mol): 105.14
Molecular Formula: C4H11NO2
InChi Key: GIAFURWZWWWBQT-UHFFFAOYSA-N
SMILES: NCCOCCO
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)