accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 929-37-3

IUPAC Name: 2-[2-(ethenyloxy)ethoxy]ethan-1-ol
Synonyms: 2-[2-(ethenyloxy)ethoxy]ethanol
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: WULAHPYSGCVQHM-UHFFFAOYSA-N
SMILES: OCCOCCOC=C
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)