accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 929-59-9

IUPAC Name: 2-[2-(2-aminoethoxy)ethoxy]ethan-1-amine
Synonyms: 2-[2-(2-aminoethoxy)ethoxy]ethanamine
Molecular Weight (g/mol): 148.21
Molecular Formula: C6H16N2O2
InChi Key: IWBOPFCKHIJFMS-UHFFFAOYSA-N
SMILES: NCCOCCOCCN
Boiling Point 
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)