accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 929-98-6

IUPAC Name: 1-(nonylsulfanyl)nonane
Synonyms: 1-(nonylsulfanyl)nonane
Molecular Weight (g/mol): 286.56
Molecular Formula: C18H38S
InChi Key: KMKSVAGOBVUFRO-UHFFFAOYSA-N
SMILES: CCCCCCCCCSCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results