Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonyl Sulfide 97.0+%, TCI America™
SDP

Catalog No. N036025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N036025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N036025G Supplier TCI America Supplier No. N036025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 929-98-6
Molecular Formula C18H38S
Molecular Weight (g/mol) 286.56
MDL Number MFCD00027342
InChI Key KMKSVAGOBVUFRO-UHFFFAOYSA-N
Synonym dinonyl sulfide, di-n-nonyl sulfide, nonyl sulfide, 1-nonylsulfanyl nonane, nonylthiononane, dinonylsulfide, n-nonylsulfide, dinonyl sulphide, n-nonyl sulfide, octylmethylsulfide
PubChem CID 525006
IUPAC Name 1-(nonylsulfanyl)nonane
SMILES CCCCCCCCCSCCCCCCCCC

Specifications

Color Yellow
Boiling Point 202°C
Quantity 25 g
Formula Weight 286.56
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonyl Sulfide
EINECSNumber (2)-0473&(2)-0475
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.