accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 929896-22-0

IUPAC Name: 2-{2,4-dimethyl-5-[4-(propan-2-yl)-4,5-dihydro-1,3-oxazol-2-yl]phenyl}-4-(propan-2-yl)-4,5-dihydro-1,3-oxazole
Synonyms: 4-isopropyl-2-[5-(4-isopropyl-4,5-dihydro-1,3-oxazol-2-yl)-2,4-dimethylphenyl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 328.46
Molecular Formula: C20H28N2O2
InChi Key: WRIRILQZUGPIMT-UHFFFAOYNA-N
SMILES: CC(C)C1COC(=N1)C1=CC(C2=NC(CO2)C(C)C)=C(C)C=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results