Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-4,6-Bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene 93.0+%, TCI America™
SDP

Catalog No. B419620MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B419620MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B419620MG Supplier TCI America Supplier No. B419620MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 929896-22-0
Molecular Formula C20H28N2O2
Molecular Weight (g/mol) 328.456
InChI Key WRIRILQZUGPIMT-QZTJIDSGSA-N
Synonym (S,S)-2,2′C-(4,6-m-Xylenediyl)bis(4-isopropyl-2-oxazoline)
PubChem CID 24744076
IUPAC Name (4S)-2-[2,4-dimethyl-5-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]phenyl]-4-propan-2-yl-4,5-dihydro-1,3-oxazole
SMILES CC1=CC(=C(C=C1C2=NC(CO2)C(C)C)C3=NC(CO3)C(C)C)C

Specifications

Melting Point 37°C
Color White
Quantity 20 mg
Formula Weight 328.46
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (S,S)-4,6-Bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.