accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOCH3

CAS RN 93-03-8

IUPAC Name: (3,4-dimethoxyphenyl)methanol
Synonyms: veratryl alcohol dimethoxybenzyl alcohol veratrole alcohol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: OEGPRYNGFWGMMV-UHFFFAOYSA-N
SMILES: COC1=CC=C(CO)C=C1OC
Boiling Point 
  • (1)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (1)
  • (5)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (6)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (4)
  • (1)
  • (5)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)