Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dimethoxybenzyl alcohol

Catalog No. D13300025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13300025G 25 g
D133000100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13300025G Supplier Sigma Aldrich Supplier No. D13300025G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-dimethoxybenzyl alcohol is widely used in the synthesis of various cyclotriveratrylenes (ctvs), which are cyclic molecular hosts with a cavity to accommodate guest molecules
  • It can also be used as a precursor in the total synthesis of salvianolic acid n

Specifications

CAS 93-03-8
Density 1.157 g/mL (at 25°C (literature))
Boiling Point 296°C to 297°C (732 mmHg)
Molecular Formula C9H12O3
Refractive Index n20/D 1.552 (literature)
Linear Formula (CH3O)2C6H3CH2OH
MDL Number MFCD00004638
Quantity 25 g
Synonym (3,4-Dimethoxyphenyl)methanol; Veratryl alcohol
Molecular Weight (g/mol) 168.19
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.