accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OHO

CAS RN 93-07-2

IUPAC Name: 3,4-dimethoxybenzoic acid
Synonyms: veratric acid veratrumenoic acid dimethylprotocatechuic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: DAUAQNGYDSHRET-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C(O)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (3)
Physical Form 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (6)
  • (3)
  • (5)
  • (2)