accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-16-3

IUPAC Name: 1,2-dimethoxy-4-[(1E)-prop-1-en-1-yl]benzene
Synonyms: methyl isoeugenol 4-trans-propenylveratrole methylisoeugenol trans-methylisoeugenol trans-isomethyleugenol
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: NNWHUJCUHAELCL-SNAWJCMRSA-N
SMILES: COC1=CC=C(\C=C\C)C=C1OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)