accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-44-7

IUPAC Name: naphthalen-2-yl benzoate
Synonyms: 2-naphthalenol, benzoate
Molecular Weight (g/mol): 248.28
Molecular Formula: C17H12O2
InChi Key: DWJIJRSTYFPKGD-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=C2C=CC=CC2=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results