Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthyl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B110825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B110825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B110825G Supplier TCI America Supplier No. B110825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93-44-7
Molecular Formula C17H12O2
Molecular Weight (g/mol) 248.281
MDL Number MFCD00014320
InChI Key DWJIJRSTYFPKGD-UHFFFAOYSA-N
Synonym 2-naphthyl benzoate, benzonaphthol, 2-naphthalenol, benzoate, betabenzon, 2-benzoyloxynaphthalene, 2-naphthylbenzoate, 2-naphthol benzoate, beta-naphthol benzoate, beta-naphthyl benzoate, benzoic acid 2-naphthyl ester
PubChem CID 66734
IUPAC Name naphthalen-2-yl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 109°C
Quantity 25 g
Formula Weight 248.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Benzoate
EINECSNumber (4)-0358
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.