accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-55-0

IUPAC Name: 1-phenylpropan-1-one
Synonyms: propiophenone propionphenone
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: KRIOVPPHQSLHCZ-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (2)
  • (6)
  • (6)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)