Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenyl Ketone, MP Biomedicals

Catalog No. ICN208876 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208876 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208876 Supplier MP Biomedicals, Inc Supplier No. 0520887680

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93-55-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
InChI Key KRIOVPPHQSLHCZ-UHFFFAOYSA-N
Synonym propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
PubChem CID 7148
ChEBI CHEBI:425902
IUPAC Name 1-phenylpropan-1-one
SMILES CCC(=O)C1=CC=CC=C1

Specifications

Quantity 100 g
Formula Weight 134.18
Chemical Name or Material Ethyl Phenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.