accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClO

CAS RN 93-67-4

IUPAC Name: 5-chloro-2-phenoxyaniline
Synonyms: 5-chloro-2-phenoxyaniline
Molecular Weight (g/mol): 219.67
Molecular Formula: C12H10ClNO
InChi Key: SXEBHIMOUHBBOS-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1OC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results