Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chlorodiphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. A222225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A222225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A222225G Supplier TCI America Supplier No. A222225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93-67-4
Molecular Formula C12H10ClNO
Molecular Weight (g/mol) 219.668
MDL Number MFCD00025217
InChI Key SXEBHIMOUHBBOS-UHFFFAOYSA-N
Synonym 2-amino-4-chlorophenyl phenyl ether, benzenamine, 5-chloro-2-phenoxy, 4-chlor-2-aminodiphenylether, 2-amino-4-chlorodiphenyl ether, 5-chloro-2-phenoxy-aniline, unii-66sm796mdb, 5-chloro-2-phenoxybenzenamine, 5-chloro-2-phenoxyphenylamine, 2-amino-4-chlorodiphenylether, 4-chloro-2-aminodiphenyl ether
PubChem CID 66738
IUPAC Name 5-chloro-2-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=C(C=C(C=C2)Cl)N

Specifications

Melting Point 43°C
Boiling Point 192°C
Quantity 25 g
Formula Weight 219.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chlorodiphenyl Ether
EINECSNumber (3)-0973
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.