accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-90-3

IUPAC Name: 2-[methyl(phenyl)amino]ethan-1-ol
Synonyms: N-methylphenylethanolamine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: VIIZJXNVVJKISZ-UHFFFAOYSA-N
SMILES: CN(CCO)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)