Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(N-Methylanilino)ethanol 97.0+%, TCI America™
SDP

Catalog No. H0359500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0359500ML 500 mL
H035925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0359500ML Supplier TCI America Supplier No. H0359500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93-90-3
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00020572
InChI Key VIIZJXNVVJKISZ-UHFFFAOYSA-N
Synonym N-(2-Hydroxyethyl)-N-methylaniline, N-Methyl-N-2-ethanolaniline
PubChem CID 62340
IUPAC Name 2-(N-methylanilino)ethanol
SMILES CN(CCO)C1=CC=CC=C1

Specifications

Boiling Point 266°C
Quantity 500 mL
Formula Weight 151.21
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-(N-Methylanilino)ethanol
EINECSNumber (3)-0116
RTECSNumber KL7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.