CAS RN 931-17-9
1,2-Cyclohexanediol, cis + trans, 98%
CAS: 931-17-9 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00003861 InChI Key: PFURGBBHAOXLIO-UHFFFAOYSA-N Synonym: 1,2-cyclohexanediol,2-hydroxycyclohexanol,pyrocatechitol,1,2-dihydroxycyclohexane,brenzkatechin,1,2-benzenediol, hexahydro,brenzkatechin german,1,2-trans-cyclohexanediol,trans-1,2-dihydroxycyclohexane,1,2-cyclohexanediol, mixture of cis and trans PubChem CID: 13601 ChEBI: CHEBI:24567 IUPAC Name: cyclohexane-1,2-diol SMILES: C1CCC(C(C1)O)O
1,2-Cyclohexanediol (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 931-17-9 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00003861 InChI Key: PFURGBBHAOXLIO-UHFFFAOYSA-N Synonym: 1,2-cyclohexanediol,2-hydroxycyclohexanol,pyrocatechitol,1,2-dihydroxycyclohexane,brenzkatechin,1,2-benzenediol, hexahydro,brenzkatechin german,1,2-trans-cyclohexanediol,trans-1,2-dihydroxycyclohexane,1,2-cyclohexanediol, mixture of cis and trans PubChem CID: 13601 ChEBI: CHEBI:24567 IUPAC Name: cyclohexane-1,2-diol SMILES: C1CCC(C(C1)O)O
MP Biomedicals, Inc 1,2-Cyclohexanediol, cis-trans, MP Biomedicals
CAS: 931-17-9 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 InChI Key: PFURGBBHAOXLIO-UHFFFAOYSA-N Synonym: 1,2-cyclohexanediol,2-hydroxycyclohexanol,pyrocatechitol,1,2-dihydroxycyclohexane,brenzkatechin,1,2-benzenediol, hexahydro,brenzkatechin german,1,2-trans-cyclohexanediol,trans-1,2-dihydroxycyclohexane,1,2-cyclohexanediol, mixture of cis and trans PubChem CID: 13601 ChEBI: CHEBI:24567 IUPAC Name: cyclohexane-1,2-diol SMILES: C1CCC(C(C1)O)O