Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Cyclohexanediol, cis + trans, 98%

Catalog No. AAA1857218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1857218 50 g
AAA1857209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1857218 Supplier Thermo Scientific Chemicals Supplier No. A1857218
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 931-17-9 | C6H12O2 | 116.16 g/mol

1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-17-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00003861
InChI Key PFURGBBHAOXLIO-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans
PubChem CID 13601
ChEBI CHEBI:24567
IUPAC Name cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

Melting Point 72°C to 76°C
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,2-Cyclohexanediol, cis + trans
EINECSNumber 213-229-8
RTECSNumber GV0220000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.