accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2

CAS RN 93138-55-7

IUPAC Name: 3-[(piperidin-1-yl)methyl]aniline
Synonyms: 3-(piperidin-1-ylmethyl)aniline
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: SEJNYLBEEMVJNN-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CN2CCCCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results