Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Piperidin-1-ylmethyl)aniline, ≥97%, Thermo Scientific™

Catalog No. CC45214DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45214DE 5 g
CC45214DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC45214DE Supplier Thermo Scientific Chemicals Supplier No. CC45214DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93138-55-7
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD04114502
InChI Key SEJNYLBEEMVJNN-UHFFFAOYSA-N
Synonym 3-piperidin-1-ylmethyl aniline, 3-piperidin-1-ylmethyl phenyl amine, 3-1-piperidylmethyl aniline, 3-piperidylmethyl phenylamine, 3-1-piperidinylmethyl aniline, 3-piperidylmethyl aniline, benzenamine, 3-1-piperidinylmethyl, 3-piperidinylmethyl aniline, 1-3-aminobenzyl piperidine
PubChem CID 6484331
IUPAC Name 3-(piperidin-1-ylmethyl)aniline
SMILES C1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material 3-(Piperidin-1-ylmethyl)aniline
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.