accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNN

CAS RN 932-35-4

IUPAC Name: 3-hydroxypyridine-2-carbonitrile
Synonyms: 3-hydroxypyridine-2-carbonitrile
Molecular Weight (g/mol): 120.11
Molecular Formula: C6H4N2O
InChi Key: XTVFTOVNAKNVQK-UHFFFAOYSA-N
SMILES: OC1=CC=CN=C1C#N
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results