Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-3-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. C16645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16645G 5 g
C166425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16645G Supplier TCI America Supplier No. C16645G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 932-35-4
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
MDL Number MFCD01646135
InChI Key XTVFTOVNAKNVQK-UHFFFAOYSA-N
Synonym 3-Hydroxypicolinonitrile
PubChem CID 101920
IUPAC Name 3-hydroxypyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)O

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 120.11
Percent Purity ≥98.0% (GC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-3-hydroxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.