accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 933-78-8

IUPAC Name: 2,3,5-trichlorophenol
Synonyms: 2,3,5-trichlorophenol
Molecular Weight (g/mol): 197.44
Molecular Formula: C6H3Cl3O
InChi Key: WWGQHTJIFOQAOC-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results