Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trichlorophenol 99%, Thermo Scientific™

Catalog No. AC168180010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC168180010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC168180010 Supplier Thermo Scientific Chemicals Supplier No. 168180010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933-78-8
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.439
InChI Key WWGQHTJIFOQAOC-UHFFFAOYSA-N
Synonym phenol, 2,3,5-trichloro, unii-6t7duc2c8b, ccris 6109, 2,3,5-tcp solution, 6t7duc2c8b, solution, pubchem12940, 2,3,5-trichlorphenol, dsstox_cid_6208, phenol,2,3,5-trichloro
PubChem CID 13619
IUPAC Name 2,3,5-trichlorophenol
SMILES C1=C(C=C(C(=C1Cl)Cl)O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 57°C to 60°C
Color White
Infrared Spectrum Authentic
Linear Formula Cl3C6H2OH
Quantity 1 g
Beilstein 06, II, 180
Formula Weight 197.45
Percent Purity ≥98.5%
Physical Form Chunks
Chemical Name or Material 2, 3, 5-Trichlorophenol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.