accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 934-90-7

IUPAC Name: 1-[(2S)-2-hydroxypropyl]piperidin-1-ium
Synonyms: 1-[(2S)-2-hydroxypropyl]piperidin-1-ium
Molecular Weight (g/mol): 144.24
Molecular Formula: C8H18NO
InChi Key: UCASFSAKVJTSET-QMMMGPOBSA-O
SMILES: C[C@H](O)C[NH+]1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results