Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Methyl-1-piperidineethanol 97%, Thermo Scientific™

Catalog No. AC255730250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC255730250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255730250 Supplier Thermo Scientific Chemicals Supplier No. 255730250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-90-7
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
InChI Key UCASFSAKVJTSET-UHFFFAOYSA-N
Synonym 1-piperidin-1-yl propan-2-ol, alpha-methyl-1-piperidineethanol, 1-piperidylpropan-2-ol, 1-piperidino-2-propanol, 1-a-piperidyl-propan-2-ol, 1-piperidin-1-yl-propan-2-ol, alpha-methyl-1-piperidine ethanol
PubChem CID 233605
IUPAC Name 1-piperidin-1-ylpropan-2-ol
SMILES CC(CN1CCCCC1)O

Specifications

Color Orange-Yellow
Density 0.93
Infrared Spectrum Authentic
Refractive Index 1.4604 to 1.4624 (20°C, 589nm)
Quantity 25 mL
Formula Weight 143.22
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material α-Methyl-1-piperidineethanol, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.