accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 934545-80-9

NBr

CAS RN 934545-80-9

IUPAC Name: 3-bromo-9-(naphthalen-2-yl)-9H-carbazole
Synonyms: 3-bromo-9-(naphthalen-2-yl)carbazole
Molecular Weight (g/mol): 372.27
Molecular Formula: C22H14BrN
InChi Key: MBGUQKKYEOAENA-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)