accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 934570-41-9

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 6-phenylpyridine-3-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 6-phenylpyridine-3-carboxylate
Molecular Weight (g/mol): 365.26
Molecular Formula: C18H8F5NO2
InChi Key: YUPUNKGUYBKZST-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CC=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results