accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 934570-50-0

IUPAC Name: 4-(3-bromothiophen-2-yl)benzoyl chloride
Synonyms: 4-(3-bromothiophen-2-yl)benzoyl chloride
Molecular Weight (g/mol): 301.58
Molecular Formula: C11H6BrClOS
InChi Key: FNHRRPMNJHHELO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results