accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONSCl

CAS RN 934570-60-2

IUPAC Name: tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
Synonyms: tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 219.68
Molecular Formula: C8H10ClNO2S
InChi Key: SQZXVWDARSIXTM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CN=C(Cl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results