Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-chloro-1,3-thiazole-5-carboxylate, 95%, Thermo Scientific™

Catalog No. MO08181DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO08181DE 5 g Amber Glass Bottle
MO08181DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO08181DE Supplier Thermo Scientific Chemicals Supplier No. MO08181DE
Add to Cart

Chemical Identifiers

CAS 934570-60-2
Molecular Formula C8H10ClNO2S
Molecular Weight (g/mol) 219.683
MDL Number MFCD09879991
InChI Key SQZXVWDARSIXTM-UHFFFAOYSA-N
Synonym tert-butyl 2-chlorothiazole-5-carboxylate, t-butyl-2-chloro-1,3-thiazole-5-carboxylate
PubChem CID 24229786
IUPAC Name tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
SMILES CC(C)(C)OC(=O)C1=CN=C(S1)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 219.69
Percent Purity 95%
Chemical Name or Material tert-Butyl 2-chloro-1, 3-thiazole-5-carboxylate
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.