accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93471-41-1

CH3SOOOCH3NHCH3

CAS RN 93471-41-1

IUPAC Name: 2,6-dimethylpyridin-1-ium 4-methylbenzene-1-sulfonate
Synonyms: 2,6-dimethylpyridin-1-ium tosylate
Molecular Weight (g/mol): 279.35
Molecular Formula: C14H17NO3S
InChi Key: ZXFCGHQVBSUJLX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=[NH+]1.CC1=CC=C(C=C1)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results