accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 936-52-7

IUPAC Name: 4-(cyclopent-1-en-1-yl)morpholine
Synonyms: 4-(cyclopent-1-en-1-yl)morpholine
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: VAPOFMGACKUWCI-UHFFFAOYSA-N
SMILES: C1CC=C(C1)N1CCOCC1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)