Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Cyclopenten-1-yl)morpholine, 96%, Thermo Scientific™

Catalog No. AAL0026222
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0026222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0026222 Supplier Thermo Scientific Chemicals Supplier No. L0026222

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-52-7
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.225
MDL Number MFCD00006162
InChI Key VAPOFMGACKUWCI-UHFFFAOYSA-N
Synonym 1-morpholinocyclopentene, n-1-cyclopenten-1-yl morpholine, 1-morpholino-1-cyclopentene, morpholine, 4-1-cyclopenten-1-yl, 4-1-cyclopenten-1-yl morpholine, 4-1-cyclopentenyl morpholine, 1-n-morpholino cyclopentene, n-1-cyclopenten-1-yl-morpholine, 4-cyclopent-1-en-1-yl morpholine
PubChem CID 70294
IUPAC Name 4-(cyclopenten-1-yl)morpholine
SMILES C1CC=C(C1)N2CCOCC2

Specifications

Density 0.957
Boiling Point 105°C to 106°C (12 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.5118
Quantity 100 g
Beilstein 117154
Formula Weight 153.23
Percent Purity 96%
Chemical Name or Material 4-(1-Cyclopenten-1-yl)morpholine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 213-316-0
RTECSNumber QE0685000
TSCA Yes
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.