accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 936-72-1

IUPAC Name: 1-methyl-2-(prop-2-en-1-yloxy)benzene
Synonyms: 1-methyl-2-(prop-2-en-1-yloxy)benzene
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: LQOGVESOGIHDSO-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results