accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 937-39-3

IUPAC Name: 2-phenylacetohydrazide
Synonyms: 2-phenylacetohydrazide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: FPTCVTJCJMVIDV-UHFFFAOYSA-N
SMILES: NNC(=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)