Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetic acid hydrazide, 98%, Thermo Scientific Chemicals

Catalog No. AC130290250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130290250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC130290250 Supplier Thermo Scientific Chemicals Supplier No. 130290250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 937-39-3 | Molecular Formula: C8H10N2O | Molecular Weight: 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-39-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007612
InChI Key FPTCVTJCJMVIDV-UHFFFAOYSA-N
Synonym phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide
PubChem CID 70301
IUPAC Name 2-phenylacetohydrazide
SMILES C1=CC=C(C=C1)CC(=O)NN

Specifications

Melting Point 115.0°C to 117.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CONHNH2
Quantity 25 g
Beilstein 09, 446
Formula Weight 150.18
Percent Purity 98%
Physical Form Needles
Chemical Name or Material Phenylacetic acid hydrazide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
EINECSNumber 213-328-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.