accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93703-24-3

IUPAC Name: 8-bromo-3-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 8-bromo-3-methyl-1,7-dihydropurine-2,6-dione
Molecular Weight (g/mol): 245.04
Molecular Formula: C6H5BrN4O2
InChi Key: QTEQVEJOXGBDGI-UHFFFAOYSA-N
SMILES: CN1C2=C(NC(Br)=N2)C(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results