Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromo-3-methylxanthine 98.0+%, TCI America™
SDP

Catalog No. B41341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41341G Supplier TCI America Supplier No. B41341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93703-24-3
Molecular Formula C6H5BrN4O2
Molecular Weight (g/mol) 245.036
MDL Number MFCD17676400
InChI Key QTEQVEJOXGBDGI-UHFFFAOYSA-N
PubChem CID 1548797
IUPAC Name 8-bromo-3-methyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)NC1=O)NC(=N2)Br

Specifications

Melting Point 300°C
Color White
Quantity 1 g
Formula Weight 245.04
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromo-3-methylxanthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.