accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSSOHOO

CAS RN 93718-83-3

IUPAC Name: 2,3-bis[(2-hydroxyethyl)sulfanyl]-1,4-dihydronaphthalene-1,4-dione
Synonyms: 2,3-bis[(2-hydroxyethyl)sulfanyl]naphthalene-1,4-dione
Molecular Weight (g/mol): 310.38
Molecular Formula: C14H14O4S2
InChi Key: MAASHDQFQDDECQ-UHFFFAOYSA-N
SMILES: OCCSC1=C(SCCO)C(=O)C2=CC=CC=C2C1=O
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results