accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrNCO

CAS RN 937795-88-5

IUPAC Name: 3-bromo-4-isocyanatothiophene
Synonyms: 3-bromo-4-isocyanatothiophene
Molecular Weight (g/mol): 204.04
Molecular Formula: C5H2BrNOS
InChi Key: HVJDEHSHJHNENX-UHFFFAOYSA-N
SMILES: BrC1=CSC=C1N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results