accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNHO

CAS RN 937795-91-0

IUPAC Name: [1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanol
Synonyms: [1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanol
Molecular Weight (g/mol): 207.28
Molecular Formula: C11H17N3O
InChi Key: OWOMYFSSKZGDBC-UHFFFAOYNA-N
SMILES: CC1=CN=CC(=N1)N1CCCC(CO)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results