Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol, 95%, Thermo Scientific™

Catalog No. CC59009DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC59009DA 1 g Amber Glass Bottle
CC59009CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC59009DA Supplier Thermo Scientific Chemicals Supplier No. CC59009DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 937795-91-0
Molecular Formula C11H17N3O
Molecular Weight (g/mol) 207.277
MDL Number MFCD09879916
InChI Key OWOMYFSSKZGDBC-UHFFFAOYSA-N
Synonym 1-6-methylpyrazin-2-yl piperid-3-yl methanol, 3-hydroxymethyl-1-6-methylpyrazin-2-yl piperidine, 1-6-methylpyrazin-2-yl piperidin-3-yl methanol, 1-6-methylpyrazin-2-yl-3-piperidyl methan-1-ol
PubChem CID 24229598
IUPAC Name [1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanol
SMILES CC1=CN=CC(=N1)N2CCCC(C2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.28
Percent Purity 95%
Chemical Name or Material [1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.