accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNBrCH3

CAS RN 937796-05-9

IUPAC Name: 1-[4-(bromomethyl)phenyl]-3,5-dimethyl-1H-pyrazole
Synonyms: 1-[4-(bromomethyl)phenyl]-3,5-dimethylpyrazole
Molecular Weight (g/mol): 265.15
Molecular Formula: C12H13BrN2
InChi Key: RHOBMGYKCJRQTA-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results