accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNHH3CCH3

CAS RN 937796-07-1

IUPAC Name: {[4-(3,5-dimethyl-1H-pyrazol-1-yl)phenyl]methyl}(methyl)amine
Synonyms: {[4-(3,5-dimethylpyrazol-1-yl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 215.3
Molecular Formula: C13H17N3
InChi Key: WVGUCDIRMDWHNO-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)N1N=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results